2,2-Dimethyl-3,4-dihydro-2H-1,4-benzothiazine 97%
Supplier: Apollo Scientific
OR937874-1G
OR937874-250MG
APOSOR937874-1GEA
420
GBP
APOSOR937874-1G
APOSOR937874-250MG
2,2-Dimethyl-3,4-dihydro-2H-1,4-benzothiazine 97%
2,2-Dimethyl-3,4-dihydro-2H-1,4-benzothiazine
Formula:
C₁₀H₁₃NS MW: 179,282 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16471298 CAS Number: 93301-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...