4-Amino-6-chloro-2-methylquinoline 95%
Supplier: Apollo Scientific
Synonyms:
6-Chloro-2-methylquinolin-4-amine
OR933094-1G
OR933094-250MG
APOSOR933094-1GEA
453
GBP
APOSOR933094-1G
APOSOR933094-250MG
4-Amino-6-chloro-2-methylquinoline 95%
4-Amino-6-chloro-2-methylquinoline
Formula:
C₁₀H₉ClN₂ MW: 192,645 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09787712 CAS Number: 66735-24-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...