4-Methyltetrahydro-2H-pyran-4-carbonitrile 97%

Supplier: Apollo Scientific
Warning

OR932404-1G OR932404-250MG
APOSOR932404-1GEA 366 GBP
APOSOR932404-1G APOSOR932404-250MG
4-Methyltetrahydro-2H-pyran-4-carbonitrile 97%
4-Methyltetrahydro-2H-pyran-4-carbonitrile
Formula: C₇H₁₁NO
MW: 125,168 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13180512
CAS Number: 856255-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR