(5-Methylpyridin-3-yl)methanol 98%

Supplier: Apollo Scientific

Synonyms: 3-Hydroxymethyl-5-methylpyridine

OR932347-1G OR932347-5G
APOSOR932347-1GEA 106 GBP
APOSOR932347-1G APOSOR932347-5G
(5-Methylpyridin-3-yl)methanol 98%
(5-Methylpyridin-3-yl)methanol
Formula: C₇H₉NO
MW: 123,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236816
CAS Number: 102074-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR