4-(3-Hydroxypropyl)benzonitrile 95%
Supplier: Apollo Scientific
OR931934-1G
OR931934-250MG
APOSOR931934-1GEA
170
GBP
APOSOR931934-1G
APOSOR931934-250MG
4-(3-Hydroxypropyl)benzonitrile 95%
4-(3-Hydroxypropyl)benzonitrile
Formula:
C₁₀H₁₁NO MW: 161,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09028771 CAS Number: 83101-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...