3-Bromo-4-methyl-5-nitro-2-pyridinone 95%

Supplier: Apollo Scientific
Warning

OR930522-1G OR930522-5G
APOSOR930522-1GEA 105 GBP
APOSOR930522-1G APOSOR930522-5G
3-Bromo-4-methyl-5-nitro-2-pyridinone 95%
3-Bromo-4-methyl-5-nitro-2-pyridinone
Formula: C₆H₅BrN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD12828179
CAS Number: 1049706-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR