3,4-Dihydro-2H-pyran-2-carbaldehyde 95%

Supplier: Apollo Scientific
Warning

OR929862-10G OR929862-5G
APOSOR929862-10GEA 0 GBP
APOSOR929862-10G APOSOR929862-5G
3,4-Dihydro-2H-pyran-2-carbaldehyde 95%
3,4-Dihydro-2H-pyran-2-carbaldehyde
Formula: C₆H₈O₂
MW: 112,127 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00059170
CAS Number: 100-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR