3-Chloro-pyrido[2,3-b]pyrazine 95%
Supplier: Apollo Scientific
Synonyms:
3-Chloropyrido[2,3-b]pyrazine
OR929712-1G
OR929712-250MG
APOSOR929712-1GEA
780
GBP
APOSOR929712-1G
APOSOR929712-250MG
3-Chloro-pyrido[2,3-b]pyrazine 95%
3-Chloro-pyrido[2,3-b]pyrazine
Formula:
C₇H₄ClN₃ MW: 165,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20925262 CAS Number: 155535-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...