2-Ethyl-4-pyrimidinecarbaldehyde 95%
Supplier: Apollo Scientific
OR929268-1G
OR929268-250MG
APOSOR929268-1GEA
393
GBP
APOSOR929268-1G
APOSOR929268-250MG
2-Ethyl-4-pyrimidinecarbaldehyde 95%
2-Ethyl-4-pyrimidinecarbaldehyde
Formula:
C₇H₈N₂O Storage Temperature: Ambient |
MDL Number:
MFCD11519862 CAS Number: 1092299-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...