1-(3-Bromophenyl)ethanamine 97%
Supplier: Apollo Scientific
OR928575-1G
OR928575-500MG
APOSOR928575-1GEA
355
GBP
APOSOR928575-1G
APOSOR928575-500MG
1-(3-Bromophenyl)ethanamine 97%
1-(3-Bromophenyl)ethanamine
Formula:
C₈H₁₀BrN MW: 200,076 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07780647 CAS Number: 74877-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...