(2-Bromoethyl)oxirane 97%

Supplier: Apollo Scientific

Synonyms: 2-(2-Bromoethyl)oxirane

OR928303-1G OR928303-5G
APOSOR928303-1GEA 197 GBP
APOSOR928303-1G APOSOR928303-5G
(2-Bromoethyl)oxirane 97%
(2-Bromoethyl)oxirane
Formula: C₄H₇BrO
MW: 151,002 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040246
CAS Number: 13287-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR