Ethyl indoline-6-carboxylate 95%
Supplier: Apollo Scientific
OR928132-1G
OR928132-250MG
APOSOR928132-1GEA
150
GBP
APOSOR928132-1G
APOSOR928132-250MG
Ethyl indoline-6-carboxylate 95%
Ethyl indoline-6-carboxylate
Formula:
C₁₁H₁₃NO₂ MW: 191,226 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09954776 CAS Number: 350683-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...