Methyl 2,4-dihydroxyphenylacetate 95%
Supplier: Apollo Scientific
OR927468-1G
OR927468-250MG
APOSOR927468-1GEA
715
GBP
APOSOR927468-1G
APOSOR927468-250MG
Methyl 2,4-dihydroxyphenylacetate 95%
Methyl 2,4-dihydroxyphenylacetate
Formula:
C₉H₁₀O₄ MW: 182,173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24726515 CAS Number: 67828-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...