2-(Chloromethyl)-5-isopropyl-1,3,4-oxadiazole 95%
Supplier: Apollo Scientific
OR925768-1G
OR925768-250MG
APOSOR925768-1GEA
728
GBP
APOSOR925768-1G
APOSOR925768-250MG
2-(Chloromethyl)-5-isopropyl-1,3,4-oxadiazole 95%
2-(Chloromethyl)-5-isopropyl-1,3,4-oxadiazole
Formula:
C₆H₉ClN₂O MW: 160,601 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08059899 CAS Number: 3914-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...