4-Vinyl-2,3-dihydrobenzofuran 95%

Supplier: Apollo Scientific

OR925611-1G OR925611-250MG
APOSOR925611-1GEA 296 GBP
APOSOR925611-1G APOSOR925611-250MG
4-Vinyl-2,3-dihydrobenzofuran 95%
4-Vinyl-2,3-dihydrobenzofuran
Formula: C₁₀H₁₀O
MW: 146,186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09030659
CAS Number: 230642-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR