Methyl 2-(1H-pyrrolo[2,3-b]pyridin-3-yl)acetate 95%

Supplier: Apollo Scientific

OR924965-1G OR924965-250MG
APOSOR924965-1GEA 525 GBP
APOSOR924965-1G APOSOR924965-250MG
Methyl 2-(1H-pyrrolo[2,3-b]pyridin-3-yl)acetate 95%
Methyl 2-(1H-pyrrolo[2,3-b]pyridin-3-yl)acetate
Formula: C₁₀H₁₀N₂O₂
MW: 190,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226228
CAS Number: 169030-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR