(2R)-2-(Methylethyl)pyrrolidine 95%
Supplier: Apollo Scientific
OR924891-100MG
OR924891-1G
OR924891-250MG
APOSOR924891-100MGEA
171
GBP
APOSOR924891-100MG
APOSOR924891-1G
APOSOR924891-250MG
(2R)-2-(Methylethyl)pyrrolidine 95%
(2R)-2-(Methylethyl)pyrrolidine
Formula:
C₇H₁₅N MW: 113,201 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18690104 CAS Number: 63328-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...