2-(1,3-Dioxolan-2-yl)thiazole 95%

Supplier: Apollo Scientific

OR924361-1G OR924361-250MG
APOSOR924361-1GEA 728 GBP
APOSOR924361-1G APOSOR924361-250MG
2-(1,3-Dioxolan-2-yl)thiazole 95%
2-(1,3-Dioxolan-2-yl)thiazole
Formula: C₆H₇NO₂S
MW: 157,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06209531
CAS Number: 24295-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR