(4-Methylphenyl)(oxo)acetic acid 95%

Supplier: Apollo Scientific

Synonyms: 2-Oxo-2-(p-tolyl)acetic acid

OR922949-1G OR922949-5G
APOSOR922949-1GEA 263 GBP
APOSOR922949-1G APOSOR922949-5G
(4-Methylphenyl)(oxo)acetic acid 95%
(4-Methylphenyl)(oxo)acetic acid
Formula: C₉H₈O₃
MW: 164,158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00961859
CAS Number: 7163-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR