(R)-(+)-3-Cyclohexenecarboxylic acid 98%

Supplier: Apollo Scientific

Synonyms: (1R)-cyclohex-3-ene-1-carboxylic acid, (R)-Cyclohex-3-enecarboxylic acid

OR922857-100G OR922857-10G OR922857-25G
APOSOR922857-100GEA 971 GBP
APOSOR922857-100G APOSOR922857-10G APOSOR922857-25G
(R)-(+)-3-Cyclohexenecarboxylic acid 98%
(R)-(+)-3-Cyclohexenecarboxylic acid
Formula: C₇H₁₀O₂
MW: 126,153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275445
CAS Number: 5709-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR