[1-(2-Chloro-phenyl)-cyclopropyl]-methanol 95%

Supplier: Apollo Scientific

Synonyms: [1-(2-Chlorophenyl)cyclopropyl]methanol

OR922731-1G OR922731-250MG
APOSOR922731-1GEA 327 GBP
APOSOR922731-1G APOSOR922731-250MG
[1-(2-Chloro-phenyl)-cyclopropyl]-methanol 95%
[1-(2-Chloro-phenyl)-cyclopropyl]-methanol
Formula: C₁₀H₁₁ClO
MW: 182,647 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374449
CAS Number: 886366-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR