5-Cyclopropylisoxazole-4-carboxylic acid 95%

Supplier: Apollo Scientific

OR922620-1G OR922620-250MG
APOSOR922620-1GEA 141 GBP
APOSOR922620-1G APOSOR922620-250MG
5-Cyclopropylisoxazole-4-carboxylic acid 95%
5-Cyclopropylisoxazole-4-carboxylic acid
Formula: C₇H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD06798100
CAS Number: 124845-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR