Ethyl 1H-pyrrolo[2,3-c]pyridine-2-carboxylate 95%
Supplier: Apollo Scientific
OR922142-1G
OR922142-250MG
APOSOR922142-1GEA
178
GBP
APOSOR922142-1G
APOSOR922142-250MG
Ethyl 1H-pyrrolo[2,3-c]pyridine-2-carboxylate 95%
Ethyl 1H-pyrrolo[2,3-c]pyridine-2-carboxylate
Formula:
C₁₀H₁₀N₂O₂ MW: 190,199 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11506124 CAS Number: 24334-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...