6-Chloro-2-methoxypyridin-3-amine 95%

Supplier: Apollo Scientific

OR921234-1G OR921234-250MG
APOSOR921234-1GEA 74.8 GBP
APOSOR921234-1G APOSOR921234-250MG
6-Chloro-2-methoxypyridin-3-amine 95%
6-Chloro-2-methoxypyridin-3-amine
Formula: C₆H₇ClN₂O
MW: 158,586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09033793
CAS Number: 914222-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR