4-(Dimethylamino)phenol hydrochloride 98%

Supplier: Apollo Scientific

OR918917-10G OR918917-1G OR918917-5G
APOSOR918917-10GEA 453 GBP
APOSOR918917-10G APOSOR918917-1G APOSOR918917-5G
4-(Dimethylamino)phenol hydrochloride 98%
4-(Dimethylamino)phenol hydrochloride
Formula: C₈H₁₁NO·ClH
MW: 173,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01707511
CAS Number: 5882-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR