3'-Hydroxy-4'-methylacetophenone 98%

Supplier: Apollo Scientific

OR918759-1G OR918759-5G
APOSOR918759-1GEA 120 GBP
APOSOR918759-1G APOSOR918759-5G
3'-Hydroxy-4'-methylacetophenone 98%
3'-Hydroxy-4'-methylacetophenone
Formula: C₉H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD06658171
CAS Number: 33414-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR