4-Amino-2-isopropylanisol 95%

Supplier: Apollo Scientific

Synonyms: 3-Isopropyl-4-methoxyaniline

OR918538-1G OR918538-250MG
APOSOR918538-1GEA 182 GBP
APOSOR918538-1G APOSOR918538-250MG
4-Amino-2-isopropylanisol 95%
4-Amino-2-isopropylanisol
Formula: C₁₀H₁₅NO
MW: 165,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026403
CAS Number: 91251-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR