2-Amino-6-chloro-5-nitrotoluene 95%

Supplier: Apollo Scientific

OR918001-1G OR918001-5G
APOSOR918001-1GEA 257 GBP
APOSOR918001-1G APOSOR918001-5G
2-Amino-6-chloro-5-nitrotoluene 95%
2-Amino-6-chloro-5-nitrotoluene
Formula: C₇H₇ClN₂O₂
MW: 186,596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026958
CAS Number: 64863-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR