Ethyl 6-oxo-1,6-dihydropyridazine-3-carboxylate 96%

Supplier: Apollo Scientific

OR917961-1G OR917961-250MG
APOSOR917961-1GEA 315 GBP
APOSOR917961-1G APOSOR917961-250MG
Ethyl 6-oxo-1,6-dihydropyridazine-3-carboxylate 96%
Ethyl 6-oxo-1,6-dihydropyridazine-3-carboxylate
Formula: C₇H₈N₂O₃
MW: 168,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00160459
CAS Number: 63001-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR