(R)-1,2,3,4-Tetrahydro-quinoline-2-carboxylic acid 95%

Supplier: Apollo Scientific

OR916748-1G OR916748-250MG
APOSOR916748-1GEA 780 GBP
APOSOR916748-1G APOSOR916748-250MG
(R)-1,2,3,4-Tetrahydro-quinoline-2-carboxylic acid 95%
(R)-1,2,3,4-Tetrahydro-quinoline-2-carboxylic acid
Formula: C₁₀H₁₁NO₂
MW: 177,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115275
CAS Number: 92977-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR