Methyl 2-(oxan-4-ylidene)acetate 97%

Supplier: Apollo Scientific

OR913773-10G OR913773-1G OR913773-5G
APOSOR913773-10GEA 584 GBP
APOSOR913773-10G APOSOR913773-1G APOSOR913773-5G
Methyl 2-(oxan-4-ylidene)acetate 97%
Methyl 2-(oxan-4-ylidene)acetate
Formula: C₈H₁₂O₃
MW: 156,179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441532
CAS Number: 138302-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR