5,6-Dihydro-4h-pyran-2-carboxylic acid 95%

Supplier: Apollo Scientific

Synonyms: 3,4-Dihydro-2H-pyran-6-carboxylic acid

OR913346-1G OR913346-250MG
APOSOR913346-1GEA 394 GBP
APOSOR913346-1G APOSOR913346-250MG
5,6-Dihydro-4h-pyran-2-carboxylic acid 95%
5,6-Dihydro-4h-pyran-2-carboxylic acid
Formula: C₆H₈O₃
MW: 128,126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08437234
CAS Number: 31518-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR