(4-Methoxy-pyridin-2-yl)-methanol 98%

Supplier: Apollo Scientific

Synonyms: (4-Methoxypyridin-2-yl)methanol

OR913249-10G OR913249-1G OR913249-5G
APOSOR913249-10GEA 492 GBP
APOSOR913249-10G APOSOR913249-1G APOSOR913249-5G
(4-Methoxy-pyridin-2-yl)-methanol 98%
(4-Methoxy-pyridin-2-yl)-methanol
Formula: C₇H₉NO₂
MW: 139,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07774100
CAS Number: 16665-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR