5-tert-Butylthiophene-2-carbonitrile 95%
Supplier: Apollo Scientific
OR912989-1G
OR912989-250MG
APOSOR912989-1GEA
728
GBP
APOSOR912989-1G
APOSOR912989-250MG
5-tert-Butylthiophene-2-carbonitrile 95%
5-tert-Butylthiophene-2-carbonitrile
Formula:
C₉H₁₁NS MW: 165,255 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04110933 CAS Number: 685120-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...