5-tert-Butylthiophene-2-carboxamide 95%
Supplier: Apollo Scientific
OR912988-1G
OR912988-250MG
APOSOR912988-1GEA
584
GBP
APOSOR912988-1G
APOSOR912988-250MG
5-tert-Butylthiophene-2-carboxamide 95%
5-tert-Butylthiophene-2-carboxamide
Formula:
C₉H₁₃NOS MW: 183,271 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04110932 CAS Number: 685120-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...