3-(4-Hydroxymethylphenoxy)propionic acid 95%

Supplier: Apollo Scientific

OR912376-1G OR912376-5G
APOSOR912376-1GEA 165 GBP
APOSOR912376-1G APOSOR912376-5G
3-(4-Hydroxymethylphenoxy)propionic acid 95%
3-(4-Hydroxymethylphenoxy)propionic acid
Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00137851
CAS Number: 101366-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR