1-Chloro-3-ethoxypropane 95%

Supplier: Apollo Scientific

OR912184-1G OR912184-250MG
APOSOR912184-1GEA 728 GBP
APOSOR912184-1G APOSOR912184-250MG
1-Chloro-3-ethoxypropane 95%
1-Chloro-3-ethoxypropane
Formula: C₅H₁₁ClO
MW: 122,593 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787549
CAS Number: 36865-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR