Methyl 3-hydroxy-2-methylbenzoate 98%

Supplier: Apollo Scientific

OR912132-1G OR912132-5G
APOSOR912132-1GEA 191 GBP
APOSOR912132-1G APOSOR912132-5G
Methyl 3-hydroxy-2-methylbenzoate 98%
Methyl 3-hydroxy-2-methylbenzoate
Formula: C₉H₁₀O₃
MW: 166,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09038292
CAS Number: 55289-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR