4-Benzoylmorpholine 98%

Supplier: Apollo Scientific

Synonyms: Morpholino(phenyl)methanone

OR911978-1G OR911978-5G
APOSOR911978-1GEA 48.6 GBP
APOSOR911978-1G APOSOR911978-5G
4-Benzoylmorpholine 98%
4-Benzoylmorpholine
Formula: C₁₁H₁₃NO₂
MW: 191,226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00188446
CAS Number: 1468-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR