3,4-Dimethoxybenzyl chloride 95%

Supplier: Apollo Scientific

Synonyms: 3,4-Dimethoxybenzyl Chloride

OR911800-10G OR911800-1G OR911800-5G
APOSOR911800-10GEA 617 GBP
APOSOR911800-10G APOSOR911800-1G APOSOR911800-5G
3,4-Dimethoxybenzyl chloride 95%
3,4-Dimethoxybenzyl chloride
Formula: C₉H₁₁ClO₂
MW: 186,635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00185554
CAS Number: 7306-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR