N-(2-Chlorophenyl)glycine 98%
Supplier: Apollo Scientific
OR911248-1G
OR911248-5G
APOSOR911248-1GEA
243
GBP
APOSOR911248-1G
APOSOR911248-5G
N-(2-Chlorophenyl)glycine 98%
N-(2-Chlorophenyl)glycine
Formula:
C₈H₈ClNO₂ MW: 185,608 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03089219 CAS Number: 6961-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...