2-N-Cyclopropylamino-5-nitropyridine 97%

Supplier: Apollo Scientific

Synonyms: N-Cyclopropyl-5-nitropyridin-2-amine

OR911058-1G OR911058-5G
APOSOR911058-1GEA 197 GBP
APOSOR911058-1G APOSOR911058-5G
2-N-Cyclopropylamino-5-nitropyridine 97%
2-N-Cyclopropylamino-5-nitropyridine
Formula: C₈H₉N₃O₂
MW: 179,176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00097476
CAS Number: 246862-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR