7-Bromo-1H-benzo[d][1,2,3]triazole 95%
Supplier: Apollo Scientific
OR910218-1G
APOSOR910218-1GEA
118
GBP
APOSOR910218-1G
7-Bromo-1H-benzo[d][1,2,3]triazole 95%
7-Bromo-1H-benzo[d][1,2,3]triazole
Formula:
C₆H₄BrN₃ Storage Temperature: Ambient |
MDL Number:
MFCD11226749 CAS Number: 1064721-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...