1-(2-Morpholinoethyl)pyrazole 97%

Supplier: Apollo Scientific

OR910110-1G OR910110-5G
APOSOR910110-1GEA 197 GBP
APOSOR910110-1G APOSOR910110-5G
1-(2-Morpholinoethyl)pyrazole 97%
1-(2-Morpholinoethyl)pyrazole
Formula: C₉H₁₅N₃O
Storage Temperature: Ambient
MDL Number: MFCD12405513
CAS Number: 1111096-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR