6-Methoxy-2,4-dihydro-1,4-benzothiazin-3-one 97%
Supplier: Apollo Scientific
OR909554-1G
OR909554-250MG
APOSOR909554-1GEA
519
GBP
APOSOR909554-1G
APOSOR909554-250MG
6-Methoxy-2,4-dihydro-1,4-benzothiazin-3-one 97%
6-Methoxy-2,4-dihydro-1,4-benzothiazin-3-one
Formula:
C₉H₉NO₂S MW: 195,238 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17212483 CAS Number: 99286-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...