2-(Oxan-4-yl)acetaldehyde 95%
Supplier: Apollo Scientific
Synonyms:
2-(Tetrahydro-2H-pyran-4-yl)acetaldehyde
OR909300-10G
OR909300-1G
OR909300-5G
APOSOR909300-10GEA
860
GBP
APOSOR909300-10G
APOSOR909300-1G
APOSOR909300-5G
2-(Oxan-4-yl)acetaldehyde 95%
2-(Oxan-4-yl)acetaldehyde
Formula:
C₇H₁₂O₂ MW: 128,169 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03425265 CAS Number: 65626-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...