5-Ethyl-2,3-dihydro-1H-inden-1-one 98%
Supplier: Apollo Scientific
OR909141-1G
OR909141-5G
APOSOR909141-1GEA
197
GBP
APOSOR909141-1G
APOSOR909141-5G
5-Ethyl-2,3-dihydro-1H-inden-1-one 98%
5-Ethyl-2,3-dihydro-1H-inden-1-one
Formula:
C₁₁H₁₂O MW: 160,212 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09261011 CAS Number: 4600-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...