2-Amino-3'-bromoacetophenone hydrochloride 97%

Supplier: Apollo Scientific
Warning

OR909021-1G OR909021-5G
APOSOR909021-1GEA 237 GBP
APOSOR909021-1G APOSOR909021-5G
2-Amino-3'-bromoacetophenone hydrochloride 97%
2-Amino-3'-bromoacetophenone hydrochloride
Formula: C₈H₈BrNO·ClH
MW: 250,52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00193063
CAS Number: 61858-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR