t-Butyl methyl fumarate 97%

Supplier: Apollo Scientific

Synonyms: tert-Butyl methyl fumarate

OR908127-1G OR908127-5G
APOSOR908127-1GEA 110 GBP
APOSOR908127-1G APOSOR908127-5G
t-Butyl methyl fumarate 97%
t-Butyl methyl fumarate
Formula: C₉H₁₄O₄
MW: 186.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26383412
CAS Number: 55556-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR