Isopropyl 4-amino-1-ethyl-1H-pyrazole-3-carboxylate 95%

Supplier: Apollo Scientific

OR906400-1G OR906400-250MG
APOSOR906400-1GEA 780 GBP
APOSOR906400-1G APOSOR906400-250MG
Isopropyl 4-amino-1-ethyl-1H-pyrazole-3-carboxylate 95%
Isopropyl 4-amino-1-ethyl-1H-pyrazole-3-carboxylate
Formula: C₉H₁₅N₃O₂
MW: 197,234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04969131
CAS Number: 956234-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR